ENA432538420 Enamine

1-(2,3-dihydro-1-benzofuran-2-carbonyl)piperidin-4-amine hydrochloride

  • MDL number MFCD28139458
  • Cas Number : 1795491-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193189

Safety Information

msds https://www.enaminestore.com/catalog/EN300-193189
Availability details for
Quantity
  • Availability
  • Add to Cart