ENA432538422 Enamine

3-chloro-4-(methoxymethyl)aniline

  • MDL number MFCD16156625
  • Cas Number : 1249156-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193200

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193200
Availability details for
Quantity
  • Availability
  • Add to Cart