ENA432538423 Enamine

hexane-2-sulfonyl chloride

  • MDL number MFCD16671638
  • Cas Number : 872309-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193203

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193203
Availability details for
Quantity
  • Availability
  • Add to Cart