ENA432538427 Enamine

4-(3-chlorophenyl)-1,2,3,6-tetrahydropyridine

  • MDL number MFCD08689313
  • Cas Number : 95884-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193213

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193213
Availability details for
Quantity
  • Availability
  • Add to Cart