ENA432538430 Enamine

3-(difluoromethoxy)azetidine

  • MDL number MFCD28132440
  • Cas Number : 1403993-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-156762

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156762
Availability details for
Quantity
  • Availability
  • Add to Cart