ENA432538431 Enamine

tert-butyl decahydro-1H-pyrido[3,2-c]azepine-6-carboxylate

  • MDL number MFCD28139461
  • Cas Number : 1784016-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193217

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193217
Availability details for
Quantity
  • Availability
  • Add to Cart