ENA432538434 Enamine

2-(4-bromophenyl)-1,1-difluoropropan-2-ol

  • MDL number MFCD23943403
  • Cas Number : 887781-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193221

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193221
Availability details for
Quantity
  • Availability
  • Add to Cart