ENA432538436 Enamine

1-(difluoromethyl)cycloheptan-1-ol

  • MDL number MFCD23943392
  • Cas Number : 1504324-80-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193223
Availability details for
Quantity
  • Availability
  • Add to Cart