ENA432538441 Enamine

4-(butan-2-yloxy)cyclohexane-1-carboxylic acid

  • MDL number MFCD28139463
  • Cas Number : 1600605-61-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-193231
Availability details for
Quantity
  • Availability
  • Add to Cart