ENA432538442 Enamine

1-(2-methoxyphenyl)-1H-pyrazole-5-carbaldehyde

  • MDL number MFCD22549375
  • Cas Number : 1159830-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193234

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193234
Availability details for
Quantity
  • Availability
  • Add to Cart