ENA432538443 Enamine

1-(2-methylphenyl)-1H-pyrazole-5-carbaldehyde

  • MDL number MFCD11975101
  • Cas Number : 1159830-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193235

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193235
Availability details for
Quantity
  • Availability
  • Add to Cart