ENA432538445 Enamine

1-(4-fluorophenyl)-1H-pyrazole-5-carbaldehyde

  • MDL number MFCD20275384
  • Cas Number : 1269291-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193237

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193237
Availability details for
Quantity
  • Availability
  • Add to Cart