ENA432538448 Enamine

3-(difluoromethyl)-1-(2-hydroxyethyl)-1H-pyrazol-5-ol

  • MDL number MFCD21667110
  • Cas Number : 1565546-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193246

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193246
Availability details for
Quantity
  • Availability
  • Add to Cart