ENA432538449 Enamine

7-fluoro-1-oxo-3,4-dihydro-1H-2-benzopyran-3-carboxylic acid

  • MDL number MFCD26381482
  • Cas Number : 1601177-71-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193247

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193247
Availability details for
Quantity
  • Availability
  • Add to Cart