ENA432538455 Enamine

1-(3-ethoxy-4-nitrophenyl)piperazine

  • MDL number MFCD18605294
  • Cas Number : 1089280-28-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193263

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193263
Availability details for
Quantity
  • Availability
  • Add to Cart