ENA432538470 Enamine

1-(2,5-difluorophenyl)-2-methylpropan-1-one

  • MDL number MFCD11205127
  • Cas Number : 1094292-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193335

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193335
Availability details for
Quantity
  • Availability
  • Add to Cart