ENA432538479 Enamine

3,6-dichloro-1H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD22553170
  • Cas Number : 1352893-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193372

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193372
Availability details for
Quantity
  • Availability
  • Add to Cart