ENA432538482 Enamine

1-(2-chloropyridin-4-yl)piperazine dihydrochloride

  • MDL number MFCD16990550
  • Cas Number : 1279029-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193380

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193380
Availability details for
Quantity
  • Availability
  • Add to Cart