ENA432538485 Enamine

5-(1H-imidazol-1-yl)pyridine-2-carboxylic acid

  • MDL number MFCD20646234
  • Cas Number : 1301214-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193383

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193383
Availability details for
Quantity
  • Availability
  • Add to Cart