ENA432538489 Enamine

2-(3-fluorophenyl)pyridin-4-ol

  • MDL number MFCD18323320
  • Cas Number : 1261904-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193389

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193389
Availability details for
Quantity
  • Availability
  • Add to Cart