ENA432538499 Enamine

(1S)-7-fluoro-1,2,3,4-tetrahydronaphthalen-1-ol

  • MDL number MFCD16037774
  • Cas Number : 1083302-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193413

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193413
Availability details for
Quantity
  • Availability
  • Add to Cart