ENA432538500 Enamine

(1R)-7-fluoro-1,2,3,4-tetrahydronaphthalen-1-ol

  • MDL number MFCD16037760
  • Cas Number : 748795-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193414

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193414
Availability details for
Quantity
  • Availability
  • Add to Cart