ENA432538502 Enamine

7-bromo-3H,4H-pyrrolo[2,1-f][1,2,4]triazin-4-one

  • MDL number MFCD27986917
  • Cas Number : 1370007-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193416

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193416
Availability details for
Quantity
  • Availability
  • Add to Cart