ENA432538503 Enamine

methyl 1,3-thiazolidine-4-carboxylate hydrochloride

  • MDL number MFCD03131299
  • Cas Number : 50703-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169-170
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193418

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193418
Availability details for
Quantity
  • Availability
  • Add to Cart