ENA432538509 Enamine

[5-chloro-2-(cyclopropylmethoxy)phenyl]methanol

  • MDL number MFCD12138901
  • Cas Number : 1154338-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193424

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193424
Availability details for
Quantity
  • Availability
  • Add to Cart