ENA432538512 Enamine

4-chloro-2-ethynyl-1-methoxybenzene

  • MDL number MFCD11618083
  • Cas Number : 95186-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193445

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193445
Availability details for
Quantity
  • Availability
  • Add to Cart