ENA432538513 Enamine

[(4-methoxyphenyl)methyl][2-(pyridin-2-yl)ethyl]amine

  • MDL number MFCD11143780
  • Cas Number : 22540-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193446

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193446
Availability details for
Quantity
  • Availability
  • Add to Cart