ENA432538514 Enamine

4-(3-aminophenyl)morpholine-3,5-dione

  • MDL number MFCD14620503
  • Cas Number : 1247823-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193447

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193447
Availability details for
Quantity
  • Availability
  • Add to Cart