ENA432538525 Enamine

methyl 2-amino-7-oxo-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

  • MDL number MFCD01855959
  • Cas Number : 124053-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193469

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193469
Availability details for
Quantity
  • Availability
  • Add to Cart