ENA432538526 Enamine

1-(3-chlorophenyl)ethane-1-sulfonamide

  • MDL number MFCD16671980
  • Cas Number : 1250227-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-104268

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104268
Availability details for
Quantity
  • Availability
  • Add to Cart