ENA432538530 Enamine

2-methyl-2-(oxan-4-yl)propanoic acid

  • MDL number MFCD18374731
  • Cas Number : 861444-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-91
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-104341

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104341
Availability details for
Quantity
  • Availability
  • Add to Cart