ENA432538536 Enamine

5-tert-butyl-2-chloropyridine

  • MDL number MFCD28040569
  • Cas Number : 102236-19-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105357

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105357
Availability details for
Quantity
  • Availability
  • Add to Cart