ENA432538538 Enamine

tert-butyl N-[4-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)phenyl]carbamate

  • MDL number MFCD24468830
  • Cas Number : 120642-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193517

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193517
Availability details for
Quantity
  • Availability
  • Add to Cart