ENA432538542 Enamine

ethyl 1-benzyl-5-oxo-4,5-dihydro-1H-pyrazole-4-carboxylate

  • MDL number MFCD25849384
  • Cas Number : 1798769-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193525

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193525
Availability details for
Quantity
  • Availability
  • Add to Cart