ENA432538543 Enamine

5-(1,1-dimethoxyethyl)-3-methyl-1,2,4-oxadiazole

  • MDL number MFCD28139344
  • Cas Number : 1202769-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105602

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105602
Availability details for
Quantity
  • Availability
  • Add to Cart