ENA432538545 Enamine

benzyl N-[(piperidin-3-yl)methyl]carbamate hydrochloride

  • MDL number MFCD11101380
  • Cas Number : 676621-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193530

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193530
Availability details for
Quantity
  • Availability
  • Add to Cart