ENA432538553 Enamine

3-(methylamino)-3-phenylpropanamide hydrochloride

  • MDL number MFCD28139496
  • Cas Number : 857770-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193595

Safety Information

msds https://www.enaminestore.com/catalog/EN300-193595
Availability details for
Quantity
  • Availability
  • Add to Cart