ENA432538558 Enamine

4,6-dichlorobenzene-1,3-dithiol

  • MDL number MFCD22042713
  • Cas Number : 60822-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-159814

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-159814
Availability details for
Quantity
  • Availability
  • Add to Cart