ENA432538559 Enamine

4-(dimethylamino)-2-(trifluoromethyl)benzaldehyde

  • MDL number MFCD23130686
  • Cas Number : 63333-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193603

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193603
Availability details for
Quantity
  • Availability
  • Add to Cart