ENA432538574 Enamine

5-bromo-4-(tert-butoxy)-2-chloropyrimidine

  • MDL number MFCD08690696
  • Cas Number : 57054-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193630

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193630
Availability details for
Quantity
  • Availability
  • Add to Cart