ENA432538576 Enamine

1,1,1-trifluoro-5-isothiocyanatopentane

  • MDL number MFCD24224697
  • Cas Number : 1549699-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-193637
Availability details for
Quantity
  • Availability
  • Add to Cart