ENA432538579 Enamine

1-(5-bromopyridin-3-yl)cyclopropane-1-carboxylic acid

  • MDL number MFCD18249881
  • Cas Number : 1256038-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193648

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193648
Availability details for
Quantity
  • Availability
  • Add to Cart