ENA432538580 Enamine

[(2-chlorophenyl)amino]formonitrile

  • MDL number MFCD12402219
  • Cas Number : 45765-25-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193649

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193649
Availability details for
Quantity
  • Availability
  • Add to Cart