ENA432538581 Enamine

N1-(cyclopropylmethyl)-4-methoxybenzene-1,2-diamine

  • MDL number MFCD20669254
  • Cas Number : 1257327-82-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193651

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193651
Availability details for
Quantity
  • Availability
  • Add to Cart