ENA432538587 Enamine

3-(4-fluorophenyl)-3-methylpyrrolidine

  • MDL number MFCD16737805
  • Cas Number : 1248610-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-170174

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170174
Availability details for
Quantity
  • Availability
  • Add to Cart