ENA432538590 Enamine

3-amino-5-methyl-1,2-oxazole-4-sulfonamide

  • MDL number MFCD28145244
  • Cas Number : 1803591-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193672

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193672
Availability details for
Quantity
  • Availability
  • Add to Cart