ENA432538596 Enamine

1-(4-fluorophenyl)-4,6-dioxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid

  • MDL number MFCD28145246
  • Cas Number : 1803600-92-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193709

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193709
Availability details for
Quantity
  • Availability
  • Add to Cart