ENA432538604 Enamine

methyl 2-({[(tert-butoxy)carbonyl]amino}amino)-2-phenylacetate

  • MDL number MFCD24536491
  • Cas Number : 182887-79-2
  • Molecular Weight : 280.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-193725

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193725
Availability details for
Quantity
  • Availability
  • Add to Cart