ENA432538605 Enamine

3-(4-fluorophenyl)pyrrolidine hydrochloride

  • MDL number MFCD03094755
  • Cas Number : 1029636-03-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193728

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193728
Availability details for
Quantity
  • Availability
  • Add to Cart