ENA432538607 Enamine

5-(2-chloro-4-fluorophenyl)-1,2-oxazole-4-carboxylic acid

  • MDL number MFCD24170051
  • Cas Number : 1506583-02-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193730

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193730
Availability details for
Quantity
  • Availability
  • Add to Cart